Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco™ Protector™ Storage Cabinets Cold Water Faucets
SDP

Catalog No. 16108235 Shop All Labconco Corporation Products
Click to view available options
Includes:
Rigid gooseneck, vacuum breaker

Deck mounted cold water faucet with rigid gooseneck

  • Rigid gooseneck-spout faucets feature a tall spout shape that extends high above the counter.
  • Spout angles downward, directing water straight into the sink.
  • The tallest spout shape available, ideal for ample clearance above the counter.
  • Provides maximum distance between spout and counter, perfect for filling tall vessels.
  • Vacuum breaker prevents water from flowing back into plumbing system.

Specifications

Product Type Storage Cabinet
Color Silver
For Use With (Equipment) Labconco™ fume hood
Includes Rigid gooseneck, vacuum breaker
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.