Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Laminin Antibody (LAM-89), Novus Biologicals™
SDP

Catalog No. NB600883 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600883 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600883 Supplier Novus Biologicals Supplier No. NB600883
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Laminin Monoclonal specifically detects Laminin in Human, Mouse, Porcine, Feline, Fish, Canine (Negative), Chicken (Negative), Guinea Pig (Negative), Goat (Negative), Rabbit (Negative), Reptile (Negative), Rat (Negative), Sheep (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Laminin
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone LAM-89
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:1000
Gene Alias LAMA, Laminin A chain, laminin subunit alpha-1, laminin, alpha 1, Laminin-1 subunit alpha, Laminin-3 subunit alpha, PTBHS, S-LAM alpha, S-laminin subunit alpha
Gene Symbols LAMA1
Host Species Mouse
Immunogen Laminin Antibody (LAM-89) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse.
Molecular Weight of Antigen 337 kDa
Purification Method Unpurified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Angiogenesis, Apoptosis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Neuroscience, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 284217
Test Specificity Laminin Antibody (LAM-89) specificity was determined by immunoperoxidase and immunofluorescent labeling of protease-digested, formalin-fixed, paraffin-embedded human or animal tissue sections, localizing specifically the basal membrane of blood vessels, epithelium, nerve, and muscle tissues. Localizes specifically in the basal membrane of blood vessels, epithelium, nerve and muscle fibers. Shows no cross-reaction with collagen type IV, fibronectin, vitronectin or chondroitin sulfates type A, B and C using a dot blot immunoassay. This antibody may be used for identification of laminin in various human tissues and bodily fluids.
Target Species Human, Mouse, Pig, Feline, Fish, Canine (Negative), Chicken (Negative), Guinea Pig (Negative), Rabbit (Negative), Rat (Negative), Sheep (Negative)
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.