Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 265 publications

$281.00 - $596.50

Specifications

Antigen LC3A
Concentration 1.0 mg/mL
Dilution Western Blot 2.0 ug/ml, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 33035707), Flow Cytometry reported in scientific literature (PMID 24419333), ELISA reported in scientific literature (PMID 20930550), Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:100-1:300. Use reported in scientific literature (PMID 21545732), Immunoprecipitation 20 ug/ 500 ug of lysate, Immunohistochemistry-Paraffin 1:200-1:400. Use reported in scientific literature (PMID 26571030), Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28253371), Southern Blot, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 31783118), Chromatin Immunoprecipitation (ChIP)
Applications Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002331SS
SDP
View Documents
Novus Biologicals
NB1002331SS
0.025 mL
Each for $281.00
Only null left
Add to Cart
 
NB1002331
SDP
View Documents
Novus Biologicals
NB1002331
0.1 mL
Each for $596.50
Only null left
Add to Cart
 
Description

Description

LC3A Polyclonal specifically detects LC3A in Human, Mouse, Rat, Amphibian, Canine, Fish, Plant, Zebrafish samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Southern Blot, Immunohistochemistry Whole-Mount.
Specifications

Specifications

LC3A
Western Blot 2.0 ug/ml, Simple Western 1:50, Chromatin Immunoprecipitation reported in scientific literature (PMID 33035707), Flow Cytometry reported in scientific literature (PMID 24419333), ELISA reported in scientific literature (PMID 20930550), Immunohistochemistry 1:200-1:400, Immunocytochemistry/Immunofluorescence 1:100-1:300. Use reported in scientific literature (PMID 21545732), Immunoprecipitation 20 ug/ 500 ug of lysate, Immunohistochemistry-Paraffin 1:200-1:400. Use reported in scientific literature (PMID 26571030), Immunohistochemistry-Frozen, Immunoblotting reported in scientific literature (PMID 28253371), Southern Blot, Immunohistochemistry (Whole-Mount) reported in scientific literature (PMID 31783118), Chromatin Immunoprecipitation (ChIP)
Polyclonal
Rabbit
Autophagy, Cancer, Cellular Markers, Cytoskeleton Markers, Hypoxia, Macroautophagy, Signal Transduction
PBS with 0.02% Sodium Azide
ATG8E, Autophagy-related protein LC3 A, Autophagy-related ubiquitin-like modifier LC3 A, LC3, LC3A, lc3-i, ii, MAP1A/1B light chain 3 A, MAP1ALC3MAP1A/MAP1B LC3 A, MAP1BLC3MAP1A/MAP1B light chain 3 A, microtubule-associated protein 1 light chain 3 alphaMAP1 light chain 3-like protein 1, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3A
MAP1LC3A
IgG
Affinity Purified
This antibody detects both LC3A and LC3B.
1.0 mg/mL
Western Blot, ChIP Assay, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH)
Unconjugated
RUO
Human, Mouse, Rat, Amphibian, Canine, Fish, Plant, Zebrafish
Q9GZQ8
84557
A synthetic peptide made to an internal portion of the human LC3 protein sequence (between residues 25-121). [Uniprot: Q9H492].
Primary
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.