Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3C, Rabbit anti-Rat, Polyclonal, Millipore Sigma™

Rabbit Polyclonal Antibody

$192.34 - $498.79

Specifications

Antigen LC3C
Dilution Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected LC3C in rat brain tissue sections.
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABC207550UL
View Documents
MilliporeSigma
ABC207550UL
50 μL
Each for $192.34
Only null left
Add to Cart
 
ABC2075200
View Documents
MilliporeSigma
ABC2075200UL
200 μL
Each for $498.79
Only null left
Add to Cart
 
Description

Description

Anti-LC3C, Cat. No. ABC2075, is a rabbit polyclonal antibody that detects Microtubule-associated proteins 1A/1B light chain 3C and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.

Microtubule-associated proteins 1A/1B light chain 3C (UniProt: Q9BXW4; also known as Autophagy-related protein LC3C, Autophagy-related ubiquitin-like modifier LC3C, MAP1 light chain 3-like protein 3, MAP1A/MAP1B light chain 3 C, MAP1A/MAP1B LC3 C, Microtubule-associated protein 1 light chain 3 gamma) is encoded by the MAP1LC3C gene (Gene ID: 440738) in human. LC3s (MAP1-LC3A, B and C) are structural proteins of autophagosomal membranes, widely used as biomarkers of autophagy. LC3A is mainly localized to perinuclear and nuclear regions, LC3B is evenly distributed throughout the cytoplasm and is also localized in the nucleolar regions, and LC3C is shown to be located in the cytoplasm and strongly in the nuclei. LC3C is synthesized with a propeptide (aa 127-147), which is removed in the mature form. LC3C is a ubiquitin-like modifier that plays a crucial role in antibacterial autophagy through the selective binding of CALCOCO2. It recruits all ATG8 family members to infecting bacteria such as S.Typhimurium. It is also reported to play a role in aggrephagy, the macroautophagic degradation of ubiquitinated and aggregated proteins. LC3C is recruited to damaged mitochondria where it mediates autophagosomal degradation of mitochondrial parts in a piecemeal fashion and is required for maintaining basal mitochondrial network homeostasis. It also mediates autophagosomal degradation of MTX1 protein and cooperates with p62 to deliver MTX1 to lysosomes. (Ref.: Koukourakis, KI et al. (2015). PLoS One DOI:10.1371/journal.pone.0137675; Le Guerroue´ et al., 2017, Mol. Cell 68(4); 786-796).
Specifications

Specifications

LC3C
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Rat
Microtubule-associated proteins 1A/1B light chain 3C;Autophagy-related protein LC3 C;Autophagy-related ubiquitin-like modifier LC3C;MAP1 light chain 3-like protein 3;MAP1A/MAP1B light chain 3 C;MAP1A/MAP1B LC3 C;Microtubule-associated protein 1 light chain 3 gamma
MAP1LC3C
IgG
Affinity Purified
This rabbit polyclonal antibody specifically detects LC3C.
Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution from a representative lot detected LC3C in rat brain tissue sections.
Polyclonal
Rabbit
Apoptosis & Cancer
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
NP_001004343.1
GST-tagged full-length human recombinant Microtubule-associated proteins 1A/1B light chain 3C (LC3C).
Primary
Stable for 1 year at 2-8°C from date of receipt.
∽17 kDa observed; 16.85 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.