Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lck Antibody (33D196), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB10056678W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056678W 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10056678W Supplier Novus Biologicals Supplier No. NB10056678AF405
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Lck Monoclonal specifically detects Lck in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Lck
Applications Western Blot
Classification Monoclonal
Clone 33D196
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P06239
Gene Alias EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16
Gene Symbols LCK
Host Species Mouse
Immunogen This monoclonal antibody was raised against a synthetic peptide containing amino acid residues of human lck protein.
Purification Method Protein G purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cell Cycle and Replication, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3932
Test Specificity Lck (33D196)
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.