Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Molecular Probes™ LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation

Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays
Description
- Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
- Kit has been optimized and validated for use with a UV laser flow cytometer
- Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
- Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
- If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
- LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs
Optimal brightness
- The LIVE/DEAD™ Fixable Blue Stain was selected based on its fluorescent properties to provide a bright signal when excited with a UV laser
- The blue-fluorescent reactive dye has an excitation maximum of ∼350 nm, making it ideal for use with an UV laser, and an emission of ∼450 nm
- By switching viability staining from a channel off of the blue 488 nm laser to the UV laser, a channel is freed up on the flow cytometer for other reagents that are difficult to find in colors other than FITC or PE
- By using the UV laser, there is no spectral overlap issues with other common dyes, thus eliminating the need to compensate the viability stains from other stains in the panel
- Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments

Specifications
Specifications
Cell Permeability | Impermeant |
Cell Type | Eukaryotic Cells |
Color | Blue |
Content And Storage | Contains 2 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO. Store at -20°C. |
Description | LIVE/DEAD™ Fixable Blue Dead Cell Stain Kit, for UV excitation |
Detection Method | Fluorescence |
For Use With (Application) | Viability Assay |
For Use With (Equipment) | Flow Cytometer |
Product Type | Dead Cell Stain Kit |
Dye Type | LIVE/DEAD™ Fixable Blue Dead Cell Stain |
Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction