Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit, for 405 nm excitation
Click to view available options
Quantity:
200 Assays
400 Assays
80 assays
Description
- Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
- Kit has been optimized and validated for use with a violet laser flow cytometer
- Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
- Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
- If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
- LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs
Low compensation
- The LIVE/DEAD™ Fixable Yellow Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser
- The yellow-fluorescent reactive dye has an excitation maximum of ∼405 nm, but it is excited well with the 405 nm violet laser
- It has an emission maxima of ∼570 nm, so it can be collected in the second or third channel on most violet laser flow cytometers
Specifications
Specifications
| Cell Permeability | Impermeant |
| Cell Type | Eukaryotic Cells |
| Color | Yellow |
| Content And Storage | Contains 10 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO. Store at -20°C. |
| Description | LIVE/DEAD™ Fixable Yellow Dead Cell Stain Kit, for 405 nm excitation |
| Detection Method | Fluorescence |
| For Use With (Application) | Viability Assay |
| For Use With (Equipment) | Flow Cytometer |
| Product Type | Stain |
| Dye Type | LIVE/DEAD™ Fixable Yellow Dead Cell Stain |
| Show More |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction