Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation

Catalog No. p-7076036
Encompass_Preferred
Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays

The LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a violet laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Low compensation

  • The LIVE/DEAD™ Fixable Aqua Stain was selected based on its fluorescent properties to minimize compensation between other violet dyes and dyes that excite off of the 488 nm blue laser
  • The aqua-fluorescent reactive dye has an excitation maximum of ∼375 nm, but it is excited well with the 405 nm violet laser
  • It has an emission maxima of ∼512 nm, so it can be collected in the second channel on most violet laser flow cytometers
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Aqua
Content And Storage Contains 5 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Description LIVE/DEAD™ Fixable Aqua Dead Cell Stain Kit, for 405 nm excitation
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Stain
Dye Type LIVE/DEAD™ Fixable Aqua Dead Cell Stain
Emission 526
Excitation Wavelength Range 375 nm
Form Solid
Format Tube(s)
Product Line LIVE/DEAD
Quantity 200 Assays
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.