Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LOXL1 Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
$243.00 - $552.50
Specifications
Antigen | LOXL1 |
---|---|
Clone | C1 |
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA541635
|
Invitrogen™
MA541635 |
200 μL |
Each for $552.50
|
|
|||||
PIMA543676
|
Invitrogen™
MA543676 |
20 μL |
Each for $243.00
|
|
|||||
Description
LOXL1 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.Specifications
LOXL1 | |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q08397 | |
4016 | |
Recombinant protein Lysyl Oxidase Like Protein 1. The antigen corresponds to amino acid range 369-574 of the target protein. | |
Antibody |
C1 | |
-20°C | |
Monoclonal | |
Liquid | |
IgG2a κ | |
Human | |
Loxl1 | |
cross-linking enzyme; LOL; Lox2; LOXL; LOXL1; Lysyl oxidase 2; lysyl oxidase homolog 1; lysyl oxidase like 1; lysyl oxidase-like 1; lysyl oxidase-like protein 1 | |
Loxl1 | |
Primary | |
Protein A/G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title