Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LRRC32/GARP Antibody, Novus Biologicals™
SDP

Catalog No. NBP224664SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224664SS 0.025 mg
NBP224664 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224664SS Supplier Novus Biologicals Supplier No. NBP224664SS
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

LRRC32/GARP Polyclonal specifically detects LRRC32/GARP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen LRRC32/GARP
Applications Western Blot, Flow Cytometry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1-3 ug/ml
Formulation 0.1 ml PBS with 0.05% Sodium Azide
Gene Accession No. Q14392
Gene Alias D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32
Gene Symbols LRRC32
Host Species Rabbit
Immunogen A portion of amino acids 250-300 of human GARP/LRRC32 was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2615
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chicken, cow, dog, mouse, rat and monkey, and 92% homologous in xenopus. FOXO3A is a 673 amino acid protein which is observed at approximately 95 kD in western blot.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.