Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LRRC32/GARP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
$206.50 - $426.00
Specifications
Antigen | LRRC32/GARP |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1-3 ug/ml |
Applications | Western Blot, Flow Cytometry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224664SS
![]() |
Novus Biologicals
NBP224664SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224664
![]() |
Novus Biologicals
NBP224664 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
LRRC32/GARP Polyclonal specifically detects LRRC32/GARP in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
LRRC32/GARP | |
Western Blot 0.5-1.0 ug/ml, Flow Cytometry 1-3 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
0.1 ml PBS with 0.05% Sodium Azide | |
D11S833Eleucine-rich repeat-containing protein 32, GARPGarpin, Glycoprotein A repetitions predominantgarpin, leucine rich repeat containing 32 | |
LRRC32 | |
IgG | |
Protein A purified | |
The amino acid sequence used as immunogen is 100% homologous in human (isoform CRA_a), chicken, cow, dog, mouse, rat and monkey, and 92% homologous in xenopus. FOXO3A is a 673 amino acid protein which is observed at approximately 95 kD in western blot. |
1.0 mg/mL | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q14392 | |
2615 | |
A portion of amino acids 250-300 of human GARP/LRRC32 was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title