Learn More
Thermo Scientific™ Luminaris Color HiGreen qPCR Master Mix, low ROX
Luminaris Color HiGreen and Luminaris HiGreen qPCR Master Mixes are universal ready-to-use solutions optimized for qPCR and two-step RT-qPCR.
$421.50 - $4690.00
Specifications
Concentration | 2X |
---|---|
Detection Method | SYBR |
GC-Rich PCR Performance | High |
PCR Method | qPCR |
Polymerase | Taq DNA Polymerase |
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | No. of Reactions | Price | Quantity & Availability | |||||
FERK0374
|
Thermo Scientific™
K0374 |
5000 Reactions |
Each for $4,690.00
|
|
|||||
FERK0373
|
Thermo Scientific™
K0373 |
1250 Reactions |
Each for $1,661.00
|
|
|||||
FERK0372
|
Thermo Scientific™
K0372 |
500 Reactions |
Each for $726.50
|
|
|||||
FERK0371
|
Thermo Scientific™
K0371 |
250 Reactions |
Each for $421.50
|
|
|||||
Description
Thermo Scientific™ Luminaris Color HiGreen Master Mixes are ready-to-use qPCR master mixes with color and premixed UDG optimized for SYBR Green chemistry using standard cycling protocols.
- Blue master mix and Yellow Sample Buffer for easy pipetting
- UDG in the master mix to prevent carry-over contamination
- Equal performance of blue and colorless master mixes
- Versions with premixed ROX or fluorescein for different real-time platforms
- Standard cycling protocols
- Excellent qPCR performance
- Specificity: Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
- Sensitivity: single copy detection
- Wide linear range: accurate quantification across 9 orders of magnitude
- Reproducibility and convenience: ready-to-use 2X master mix
Specifications
2X | |
High | |
Taq DNA Polymerase | |
7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P | |
qPCR Master Mix | |
2 X |
SYBR | |
qPCR | |
Standard | |
Luminaris | |
Gene Expression | |
DNA, dsDNA |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.