Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ LY 2389575 hydrochloride
Selective negative allosteric modulator of mGlu3
Specifications
Purity | >98% |
---|---|
Product Type | LY 2389575 hydrochloride |
Molecular Weight (g/mol) | 438.58 |
Inhibitors | LY 2389575 hydrochloride |
Description
Selective negative allosteric modulator of mGlu3 (IC50 = 190 nM). Exhibits > 65-fold selectivity for mGlu3 over other mGlu receptors. Abolishes the neuroprotective action of LY 379268 against amyloid β toxicity in mixed cortical neuronal and astrocyte cell cultures.Specifications
>98% | |
LY 2389575 hydrochloride | |
438.58 | |
LY 2389575 hydrochloride |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title