Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6C Antibody (ER-MP20) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7200406 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017721 0.025 mg
NB10065413 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017721 Supplier Novus Biologicals Supplier No. NB100654130.025MG
Only null left
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Ly-6C Monoclonal specifically detects Ly-6C in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen Ly-6C
Applications Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone ER-MP20
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10-1:50, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. P0CW02
Gene Alias Ly 6c, Ly6c protein, Lymphocyte antigen 6 complex, Lymphocyte antigen 6 complex locus C, Lymphocyte antigen 6C, Lymphocyte antigen Ly 6C
Gene Symbols Ly6c1
Host Species Rat
Immunogen Balb/c macrophage precursor cell hybrids
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17067
Test Specificity Ly6C (ER-MP20)
Target Species Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.