Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lysozyme Antibody (BGN/0696/5B1) - BSA Free, Novus Biologicals™
SDP

Catalog No. p-7199338 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017536 0.025 mg
NB10063062 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017536 Supplier Novus Biologicals Supplier No. NB100630620.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 3 publications

Lysozyme Monoclonal specifically detects Lysozyme in Human, Mouse samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.

Specifications

Antigen Lysozyme
Applications Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF, ELISA, Immunocytochemistry, Immunohistochemistry, Flow Cytometry
Classification Monoclonal
Clone BGN/0696/5B1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:10-1:500, CyTOF-ready
Formulation Phosphate buffered saline with 0.09% Sodium Azide
Gene Accession No. P61626
Gene Alias EC 3.2.1.17, lysozyme, lysozyme (renal amyloidosis)1,4-beta-N-acetylmuramidase C, lysozyme C, LZM, renal amyloidosis
Gene Symbols LYZ
Host Species Mouse
Immunogen Native, from urine.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4069
Test Specificity NB100-63062 recognizes lysozyme, an enzyme that is present in mucosal secretions such as saliva and tears and is involved with non-specific organism defence. It functions by hydrolyzing 1,2-beta linkages between N-acetylmuramic acid and N-acetyl-D-glucosamine residues such as those found in the walls of various Gram-positive bacteria.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.