Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns

Catalog No. 088647
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
150 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
Stationary Phase:
PSDVB (Polystyrene Divinylbenzene)
Phenyl (Ph)

Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.

Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.

Compatible with organic solvent used in HPLC and MS under RP conditions
  • Resin is based on a hydrophobic supermacroporous 4μm polymer particles
  • Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
Efficient with low carry over
  • 1,500Å large pore size polymeric resin
  • Efficient separation of protein molecules with very low carry over
Designed for reversed-phase biomolecule LC and LC/MS separations
  • Superior resolution power for monoclonal antibodies and related substances
  • High efficiency with low carry-over
  • Excellent MS compatibility
  • Wide operating pH range: 0 to 14
  • High temperature stability: up to 110°C
  • High throughput
TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 100 mm
Max. Pressure 4000 psi (275 bar)
Particle Size 4 μm
pH 0 to 14
Pore Size 1500 Å
Quantity Each
Temperature <110°C
Stationary Phase Phenyl (Ph)
Product Line MAbPac
Packing Material DVB (Divinylbenzene) Polymer
Column Type Reversed Phase
Phase Reversed Phase
Technique Bio-LC
Green Features Responsibly packaged
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.