Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva MabSelect Sure™ Chromatography Media
GSA_VA

Catalog No. 45002989 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
200 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-002-989 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-002-989 Supplier Cytiva Supplier No. 17543802
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MabSelect™ SuRe™ is a protein A affinity medium for capturing MAbs at process scale with an alkali-tolerant rProtein A ligand that allows the for cost-effective sanitization protocols

Has an alkali-tolerant rProteain A ligand designed to enhance protease resistance to produce monoclonal antibodies

  • Withstands rigorous CIP and sanitization procedures with 0.1 to 0.5 M NaOH
  • Design gives enhanced protease resistance resulting in lower ligand leakage
  • Generic elution profile for different monoclonal antibodies enables platform approach to purification
  • High-flow agarose matrix allows the processing of large feed volumes
  • Used in the commercial manufacture of monoclonal antibodies

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 mL
For Use With (Application) For Use of Rigorous and Cost-Effective CIP and Sanitization Protocols, Manufacture of Monoclonal Antibodies
Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
Storage Requirements 2°C to 8°C
Binding Capacity 30 mg Human lgG/mL (at 2.4 min. residence time)
Product Type MabSelect SuRe Media
Flow Rate <500 cm/Hoμr
Particle Size 85 μm
Height (English) Bed 7.87 in.
Height (Metric) Bed 20 cm
Includes Certificate of Analysis
Max. Pressure <200 kPa
Matrix Highly Cross-Linked Agarose
pH Range 3 to 12
Storage Buffer 20% Ethanol
Ligand Type Alkali-stabilized Protein A-derived (E. coli)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.