Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GVS™ Magna Supported Nylon Charged Transfer Membrane

Catalog No. NP0HY330F2 Shop All Osmonics Products
Change view
Click to view available options
Quantity:
25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP0HY330F2 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP0HY330F2 Supplier GVS™ Supplier No. 1226575
Only null left
Add to Cart
Add to Cart

Ideally suited for all probes both radioactive and non-radioactive, including chemiluminescent and biotinylated detection systems

  • Non-sterile
  • Positively charged modified nylon membrane, specifically designed to allow for numerous reprobings
  • Offers significantly increased binding, maximum lot-to-lot consistency, and excellent signal retention
  • Applications: 
    • Southern transfer,
    • Northern transfers
    • Multiple reprobings
    • Alkaline Blotting
    • UV Cross linking
    • Radiolabeled detection systems
    • Non-radiolabeled detection systems

Specifications

Length (Metric) 30 cm
Width (Metric) 30 cm
Description Charged nylon transfer membrane
Color White
Quantity 25
Material Nylon
Pore Size 0.45 μm
Dimensions (LxW) 30 x 30 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.