Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mannitol, Reagent, ACS, Spectrum™ Chemical
SDP

Catalog No. 18604233 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

M1120, 69-65-8, C6H14O6

Spectrum™ Chemical Mannitol, Reagent, ACS , otherwise known as Mannite, is classified as a sugar alcohol and is most commonly observed in it′s white crystalline solid form. It has several uses, ranging from inducing osmotic stress in plants to being used to produce tablets of medicine. It can also be added to boric acid to increase the strength of the acid in order to allow for a better degree of accuracy in volumetric analysis of the acid. As an ACS grade Reagent, Spectrum Chemical manufactured mannitol is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.

Chemical Identifiers

CAS 69-65-8
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064287
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

CAS Max % 100%
Loss on Drying 0.05%
Packaging Poly Pail
Quantity 4 x 2.5 kg
Specific Rotation +23.3° to +24.3°
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.