Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MAP2 Antibody, Novus Biologicals™
SDP

Catalog No. NB300213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300213 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB300213 Supplier Novus Biologicals Supplier No. NB300213
Only null left
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

MAP2 Polyclonal antibody specifically detects MAP2 in Human, Mouse, Rat, Feline samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Knockdown.

Specifications

Antigen MAP2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:10000-1:20000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/Immunofluorescence 1:5000-1:10000, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen 1:1000-1:5000, Knockdown Validated
Formulation Supplied as concentrated IgY in PBS with 5mM Sodium Azide
Gene Accession No. P11137
Gene Alias DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2
Gene Symbols MAP2
Host Species Chicken
Immunogen This MAP2 antibody was developed against a mix of recombinant human construct of projection domain sequences, amino acids 377-1505: Prot-r-MAP2-P1, Prot-r-MAP2-P2 and Prot-r-MAP2-P3.
Molecular Weight of Antigen 199 kDa
Purification Method Ammonium sulfate precipitation
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Cytoskeleton Markers, MAP Kinase Signaling, Neuronal Stem Cells, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4133
Test Specificity This antibody was raised against recombinant constructs of the entire human projection domain, and so recognizes the high molecular MAP2 forms, MAP2A and MAP2B.
Target Species Human, Mouse, Rat, Feline
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.