Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEF2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $464.00
Specifications
Antigen | MEF2B |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18021820
![]() |
Novus Biologicals
NBP18021820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180218
![]() |
Novus Biologicals
NBP180218 |
100 μL |
Each for $464.00
|
|
|||||
Description
MEF2B Polyclonal specifically detects MEF2B in Human, Mouse samples. It is validated for Western Blot.Specifications
MEF2B | |
Polyclonal | |
Purified | |
RUO | |
NP_032604 | |
4207 | |
Synthetic peptide directed towards the C terminal of human MEF2B. Peptide sequence PVARSLCKEGPPSRGASPPTPPVSIKSERLSPVTGTSGDFPRSFPYPLLL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
LOC729991-MEF2B readthrough transcript, MEF2B, myocyte enhancer factor 2B | |
MEF2BNB-MEF2B | |
IgG | |
Protein A purified | |
38 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title