Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Meiosis Manipulatives Kit
SDPSCI_ED

Catalog No. s05932 Shop All United Scientific Supplies, Inc Products
Click to view available options
Class Size:
5 Lab Groups

The Meiosis Manipulatives Kit makes the complex biological process of meiosis come alive in your classroom.

  • Teacher demonstration models with 30 large chromosomes arranged in five colorful sets.
  • Each set consists of a maternal and paternal homolog and four sister chromatids.
  • Alleles and parental lineage are clearly identified.
  • The fifth set has removable alleles for demonstrating cross-over.
  • Student models consist of smaller sets of chromosomes.
  • Kit contents are reusable.
  • Dimensions: 53.3 x 48.3 x 12.7 cm (21 x 19 x 5 in.)

Specifications

Product Type Meiosis Manipulatives Kit
Grade 5+
For Use With (Application) Student kit for learning meiosis.
Includes Downloadable resources and teacher manual with recommended student activity, assessment options and student activity sheets.
Dimensions (L x W x H) 53.3 x 48.3 x 12.7 cm (21 x 19 x 5 in.)
Class Size 5 Lab Groups
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.