Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEIS2 Antibody (PCRP-MEIS2-2B3), Novus Biologicals™
SDP

Catalog No. NB232480 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB232480 20 μg
NB233465 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB232480 Supplier Novus Biologicals Supplier No. NBP32052020UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEIS2 Monoclonal antibody specifically detects MEIS2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen MEIS2
Applications Western Blot, Flow Cytometry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-MEIS2-2B3
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS with 0.05% BSA
Gene Alias Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2
Host Species Mouse
Immunogen Recombinant fragment of human MEIS2
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Vision
Primary or Secondary Primary
Gene ID (Entrez) 4212
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.