Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanoma antigen family C2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP1689620 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP1689620 20 μL
NBP168961 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP1689620 Supplier Novus Biologicals Supplier No. NBP16896120UL

Rabbit Polyclonal Antibody

Melanoma antigen family C2 Polyclonal specifically detects Melanoma antigen family C2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Melanoma antigen family C2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS and 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9UBF1
Gene Alias Cancer/testis antigen 10, CT10MAGE-E1 antigen, HCA587MAGE-C2, hepatocellular cancer antigen 587, Hepatocellular carcinoma-associated antigen 587, MAGE-C2 antigen, MAGEE1MGC13377, melanoma antigen family C, 2, melanoma antigen, family E, 1, cancer/testis specific, melanoma-associated antigen C2
Gene Symbols MAGEC2
Host Species Rabbit
Immunogen Synthetic peptides corresponding to MAGEC2 (melanoma antigen family C, 2) The peptide sequence was selected from the N terminal of MAGEC2. Peptide sequence SCCSSFSWSSFSEESSSQKGEDTGTCQGLPDSESSFTYTLDEKVAELVEF.
Molecular Weight of Antigen 41 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51438
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.