Learn More
Melanoma Associated Antigen Antibody (MAA/1414), Alexa Fluor™ 594, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Melanoma Associated Antigen |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | MAA/1414 |
| Conjugate | Alexa Fluor 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Chronic μgcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like |
| Host Species | Mouse |
| Immunogen | proteins extracted from a melanoma |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.