Learn More
Description
Specifications
Specifications
| Antigen | MEOX 2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4F6 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | growth arrest-specific homeo box, Growth arrest-specific homeobox, homeobox protein MOX-2, mesenchyme homeo box 2 (growth arrest-specific homeo box), mesenchyme homeobox 2GAX, MOX2mesenchyme homeobox 2 (growth arrest-specific homeo box) |
| Gene Symbols | MEOX2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
