Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Metallothionein-1A Antibody (UC1MT), Novus Biologicals™
SDP

Catalog No. NBP19749302 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.05 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP19749302 0.2 mg
NB018787 0.025 mg
NBP19749305 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP19749302 Supplier Novus Biologicals Supplier No. NBP1974930.2MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 5 publications

Metallothionein-1A Monoclonal specifically detects Metallothionein-1A in Human, Mouse, Rat, Equine, Fish, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Enzyme Immunoassay.

Specifications

Antigen metallothionein
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Monoclonal
Clone UC1MT
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Enzyme Immunoassay
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Host Species Mouse
Immunogen Cross-linked rabbit liver Metallothionein I and II.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Equine, Fish, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.