Alfa Aesar™ Methylcyclohexane, 99+%

Manufacturer: Alfa Aesar™ 3186136

Quantity 500g

 View more versions of this product

Catalog No. AA3186136

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-87-2
    Formula Weight 98.19
    MDL Number MFCD00001497
    UN Number UN2296
    Beilstein 505972
    Merck Index 14,6047
    Chemical Name or Material Methylcyclohexane

    Specifications

    Formula Weight 98.19
    Health Hazard 3 GHS P Statement
    P210-P273-P243-P301+P330+P331-P403
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid release to the environment.
    Take precautionary measures against static discharge.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store in a well-ventilated place.
    Merck Index 14,6047
    Health Hazard 2 GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.
    RTECS Number GV6125000
    Quantity 500g
    Physical Form Liquid
    MDL Number MFCD00001497
    Beilstein 505972
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLCYCLOHEXANE
    Flash Point −4° (−25°F)
    Boiling Point 101°C
    UN Number UN2296
    Health Hazard 1 Danger
    Color Colorless
    Melting Point −126°C
    Refractive Index 1.4230
    TSCA TSCA
    CAS 108-87-2
    Density 0.771g/mL
    Chemical Name or Material Methylcyclohexane
    Assay ≥99%

    GHS H Statement
    H225-H304-H315-H336
    Highly flammable liquid and vapor.
    May be fatal if swallowed and enters airways.
    Causes skin irritation.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P273-P243-P301+P330+P331-P403
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Avoid release to the environment.
    Take precautionary measures against static discharge.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store in a well-ventilated place.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit