Learn More
Methylcyclohexane, 99%, Thermo Scientific Chemicals
Description
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methylcyclohexane is used as a solvent and reagent in organic synthesis and polymer chemistry. It is a component of jet fuel and also used as correction fluid in certain consumer products. It acts as a starting material for toluene synthesis
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | -126°C |
Density | 0.771 |
Boiling Point | 101°C |
Flash Point | −4°C (−25°F) |
Refractive Index | 1.423 |
Quantity | 2500 mL |
UN Number | UN2296 |
Beilstein | 505972 |
Merck Index | 14,6047 |
Solubility Information | Immiscible with water. |
Show More |
Safety and Handling
Hazard Category | H225-H304-H315-H319-H335-H336 |
Hazard Statement | GHS H Statement H225-H304-H315-H336 Highly flammable liquid and vapour. May be fatal if swallowed and enters airways. Causes skin irritation. May cause drowsiness or dizziness. |
Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHYLCYCLOHEXANE |
EINECSNumber | 203-624-3 |
RTECSNumber | GV6125000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.