Learn More
Description
Specifications
Specifications
| Antigen | MHC Class II RT1B |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), CyTOF |
| Classification | Monoclonal |
| Clone | OX-6 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:10-1:500, Immunohistochemistry Free-Floating reported in scientific literature (PMID 31317505), CyTOF-ready |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Alias | anti-OX-6 RT1B, anti-RT1B OX-6, Clone OX6, DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC class II DQ, MHC DPB1, Non-polymorphic MHC Class II, RING6, RING7, RT1B |
| Gene Symbols | RT1-B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
