Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Mica Schist Specimen
SCI_ED

Catalog No. S26654 Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm

Mica Schist Specimen for use in lessons on metamorphic rock properties, classification, and identification.

  • Distinctly altered mineral crystals are ideal for students studying metasomatism in metamorphic rocks
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Product of moderate grade metamorphism from granite
  • Has undergone moderate levels of alteration
  • Schist is good for demonstrating effect of metamorphism on various minerals

Specifications

Product Type Metamorphic Rock, Mica Schist
Quantity 1 Ea.
Specimen Size 7 to 8 cm
Weight (Metric) 0.5 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.