Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Micro Spoon, Stainless Steel
SDPSCI_ED

Catalog No. p-200009344 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Length (Metric):
16.5 cm
23.5 cm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
S16028 23.5 cm
S15826 16.5 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S16028 Supplier United Scientific™ Supplier No. SSFS09
Only null left
Add to Cart
Add to Cart

Economical option for everyday lab use.

  • Stainless steel micro spoon has an overall length of 23.5 cm (9.25 in.).
  • One end is a spoon and one end is flat.

Specifications

Sterility Non-sterile
Autoclavable Autoclavable
Product Type Spatula
Material Stainless Steel
Length (Metric) 23.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.