Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Microsart™ AMP Mycoplasma Quantitative Detection Kit
Rapid Real-Time PCR detection kit. Sartorius Microsart™ AMP Mycoplasma Quantitative Detection Kit is used for direct detection of Mollicutes in cell culture, media components and derived biologicals.
Specifications
Content And Storage | Positive control, buffer, PCR grade water. 2° to 8°C |
---|---|
Detection Method | qPCR |
Product Type | Assay kit |
Target | 16s RNA of Mycoplasma |
Sample Type | Lyophilized |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
14-577-839
|
Sartorius
SMB951001 |
25 tests | N/A | N/A | N/A | ||||
14-577-840
|
Sartorius
SMB951002 |
100 tests | N/A | N/A | N/A | ||||
Description
- Sensitive detection of Mycoplasma DNA
- Primer, probes designed against rRNA gene Mycoplasma species
- Sample type: Master cell banks, virus seed lots, virus harvests, final lot of biologicals produced by cell
- Detection limit: 10cfu/mL
- Sample volume: 200μl to 18mL
Applications
- Used in quality control analysis in microbiological and biopharmaceutical labs
Compliance
EP 2.6.7
Specifications
Positive control, buffer, PCR grade water. 2° to 8°C | |
Assay kit | |
Lyophilized |
qPCR | |
16s RNA of Mycoplasma | |
18 to 200μL |
Safety and Handling
ShelfLife : Six months
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title