Learn More
Description
- Enables fast and reliable detection of Mycoplasma DNA in cell culture supernatants
- Detects 16 S rRNA gene of at least 110 Mycoplasma species
- Time: Three hours
- Sample can be added directly to PCR reaction tube
- TaqMan™ real-time qPCR based chemistry
- Sample volume: 200μL
Applications:
- Used with any Real-time PCR instrument which detects the dyes FAM, ROX
- Used in quality control analysis in microbiological and biopharmaceutical labs
Compliance
EP 2.6.7
Specifications
Specifications
| Content And Storage | 2° to 8°C |
| Detection Method | qPCR |
| No. of Tests | 25 tests |
| Product Type | Assay test |
| Quantity | 25 tests |
| Target | 16s rRNA of Mycoplasma |
| For Use With (Application) | Microbial Detection |
| No. of Reactions | 25 reactions |
| Sample Type | Lyophilized |
| Volume | 2μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.