Learn More
Eisco™ Milky Quartz Mineral Specimen
Shop All Eisco Products
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Description
- For use in classroom identification labs and exercises
- For use in lessons on mineral hardness, luster, and fracture
- Correlates well to Next Generation Science Standards Evidence Statements
- Used in identification exercises, asking students to distinguish between it and other visually similar minerals such as feldspar or calcite
Specifications
Specifications
Product Type | Mineral, Milky Quartz |
For Use With (Application) | Lessons on mineral hardness, luster, and fracture |
Packaging | Clear labeled and barcoded bag |
Quantity | 12/Pk. |
Specimen Size | 2 to 3 cm |