Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ Miller™ Adjustable-Fit Harnesses

Specifications

Length (English) 1.75 in.
Length (Metric) 4.4 cm
View More Specs

Products 3
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
17-987-22A
View Documents
Honeywell™
84284/SYK/DEL
Tongue buckle; Small Tongue buckle; Small N/A N/A N/A
17-987-22C
View Documents
Honeywell™
84284/LYK
Tongue buckle; Large Tongue buckle; Large N/A N/A N/A
19-034-823D
View Documents
Honeywell™
8428/XLYK
Friction buckle; X-Large Friction buckle; X-Large N/A N/A N/A
Description

Description

Lightweight harnesses designed to arrest the most severe free falls. Constructed of high-visibility 1 3/4 in. (4.4cm) nylon webbing. Leg, waist, and shoulder straps adjust easily and fit comfortably. With friction waist buckle and shoulder buckle for custom fit. Sub-pelvic strap provides additional wearing comfort.

17-987-22 series has tongue-buckle leg straps for extra security; 19-034-823 has friction buckle.

Specifications

Specifications

1.75 in.
4.4 cm
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.