Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Miller™ DuraHoist™ Accessories
For use with DuraHoist Confined Space Systems
Specifications
Certifications/Compliance | OSHA, ANSI, CSA |
---|---|
For Use With (Equipment) | DuraHoist confined space system |
Load Bearing Capacity (English) | 450 lb. |
Load Bearing Capacity (Metric) | 204 kg |
Catalog Number | Mfr. No. | Product Type | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Material | Price | Quantity & Availability | ||||
19-175-447
|
Honeywell™
DH7ZP/ |
Floor Mount Sleeve | Zinc-plated Mild Steel | N/A | N/A | N/A | |||
19-175-445
|
Honeywell™
DH3/ |
One piece adjustable mast | Aluminum | N/A | N/A | N/A | |||
Description
One-Piece Adjustable Mast
- Mast offset adjusts from 12 to 29 in. (30 to 74cm)
Three-Piece Portable Base
- Lightweight aluminum components; adjustable aluminum screws and a built-in level indicator
- No tools required for assembly
Floor Mount Sleeve
- Mounts to horizontal concrete or steel structures
Wall Mount Sleeve
- Mounts to vertical concrete or steel structures
Flush Floor Mount Sleeve
- Eliminates potential trip hazard
- Installs into 4 in. dia. (10cm) core hole
Vehicle Hitch Mount Sleeve
- Installs into 2 in. dia. (5cm) receiver on an attendant vehicle
- Provides portable anchor point
Specifications
OSHA, ANSI, CSA | |
DuraHoist confined space system | |
450 lb. | |
204 kg |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title