Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Miller™ Revolution™ Arc-Rated Harnesses
Protect from falls caused by electric arc flash and arc blast exposure
Specifications
Material | Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers |
---|---|
For Use With (Application) | Designed to protect workers from falls caused by electric arc-flash and arc-blast exposure |
Catalog Number | Mfr. No. | Product Type | Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Size | Price | Quantity & Availability | ||||
17-987-606
|
Honeywell™
RKNARQC/S/MBK |
Arc-Rated Harness without Rescue Loops | Small/Medium | N/A | N/A | N/A | |||
19-169-921
|
Honeywell™
RKNARQC/UBK |
Arc-Rated Harness without Rescue Loops | Universal | N/A | N/A | N/A | |||
19-169-922
|
Honeywell™
RKNARRLQC/UBK |
Arc-Rated Harness with Rescue Loops | Universal | N/A | N/A | N/A | |||
Description
- Kevlar™/Nomex™ DualTech™ webbing has 10,000 lb. tensile strength; engineered with permanent fire-retardant properties
- Kevlar outer offers durability and fire resistance while Nomex inner gives greater comfort
- PivotLink™ design allows freedom of motion while cam buckles provide easy, two-finger adjustment
- Feature quick-connect chest and leg buckles, cam buckles, a self-contained label pack, web finials, a Kevlar web back D-ring, and an ErgoArmor™ back shield
- Leather insulators under all-metal hardware increase worker protection
- Bright red binding on every pad for easy identification
Order Info
Universal size. Harnesses are rated to 400 lb. capacity.
Certifications
Meet ASTM™ F887-05 arc-flash standard.
Compliance
Meets OSHA 1910.66 and OSHA 1926.502 (d); ANSI™ A10.32 and ANSI Z359-2007; ASTM™ F887-13; CSA Z259.10 requirements.
Specifications
Dupont™ Kevlar™ aramid fiber, Nomex™ aramid fibers | |
Designed to protect workers from falls caused by electric arc-flash and arc-blast exposure |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title