Learn More
Eisco™ Mineral ID Kit
Shop All Eisco Products
Click to view available options
Product Type:
ID Kit with 9 Minerals
ID Kit without Minerals
Description
- 7 piece kit includes seven essential tools - reusable streak plate, glass plate, dropper bottle, magnet, nail, copper square & retractable hand lens
- Portable, pocket size tools are perfect for field testing
- Comes in zip lock bag for easy storage
- Useful for demonstrations in identifying and classifying rocks and minerals
- Perfect for group activities
- Test mineral hardness using the glass plate, nail and copper square
- Test streak color by rubbing mineral across the streak plate
- Using the magnet, determine if your mineral has strong, weak or no magnetic properties
- LDPE dropper bottle useful for storing and performing an effervescence test (requires dilute hydrochloric acid, not included)
- The retractable magnifying glass is perfect for close up examinations of your minerals and testing results
- Option to purchase with 9 piece mineral kit (calcite, crystal quartz, gypsum, hematite, magnetite, microcline, muscovite, pyrite, talc)
Specifications
Specifications
| Product Type | ID Kit without Minerals |
| For Use With (Application) | Identifying and Classifying Rocks and Minerals |
| Includes | Seven Essential Tools |