Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Amplex™ Red and Amplex™ UltraRed Reagents

Catalog No. p-4924897
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
5 x 1 mg
10 x 10 mg
Dye Type:
Amplex Red Reagent
Amplex UltraRed Reagent
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Dye Type
A12222 5 mg Amplex Red Reagent
A22177 10 x 10 mg Amplex Red Reagent
A36006 5 x 1 mg Amplex UltraRed Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A12222 Supplier Invitrogen™ Supplier No. A12222
Only null left
Add to Cart
Edge
Add to Cart

The Amplex Red and UltraRed reagents are fluorogenic substrates for peroxidase. These highly sensitive and stable probes react with H2O2 in the presence of HRP for solution-based assays and ELISAs. Amplex UltraRed is a brighter version of Amplex Red with an improved pH range and sensitivity.

The Amplex Red and Amplex UltraRed reagents are highly sensitive and stable fluorescent probes for the detection of hydrogen peroxide (H2O2) and ideal fluorogenic substrates for peroxidase. These reagents can be used for solution-based assays to detect the activity of H2O2 released from biological samples or with HRP-based ELISA applications. The Amplex UltraRed reagent is an improved version of Amplex Red that is brighter and more sensitive to peroxidase with less pH sensitivity compared to the Amplex Red reagent.

In the presence of horseradish peroxidase (HRP), the Amplex Red and UltraRed reagents react in a 1:1 stoichiometry with H2O2 to produce highly fluorescent, strongly absorbing resorufin (ex/em ∼570/585 nm) or Amplex UltraRed (ex/em ∼568/581 nm), respectively. These oxidized reaction products have a broad red emission with little interference from blue and green autofluorescence found in most biological samples. Because H2O2 is produced in many different enzymatic reactions, the Amplex Red and UltraRed reagents coupled with HRP can be used to detect the activity of many different enzymes. These reagents are also used to detect H2O2 released from biological samples, including cells and isolated mitochondria.

Features of the Amplex Red reagent:

  • Sensitive and fast detection of HRP activity or H2O2 for solution-based assays and ELISAs
  • Detection of H2O2 produced in many different enzymatic reactions
  • Measurement with either fluorescence or absorbance, permitting either fluorogenic or chromogenic detection

The Amplex Red reagent has been used to develop a variety of novel fluorogenic and chromogenic assays for enzymes that produce hydrogen peroxide. When coupled with added horseradish peroxidase, these assays that generate H2O2 permit the ultrasensitive quantitation of a diverse assortment of analytes, including glucose, galactose, cholesterol, glutamic acid, xanthine (or hypoxanthine), uric acid, choline, and acetylcholine.

The Amplex UltraRed reagent offers brighter fluorescence and enhanced sensitivity on a per-mole basis in peroxidase or peroxidase-coupled enzyme assays compared to Amplex Red. The Amplex UltraRed reagent is also less sensitive to acidic pH and exhibits improved stability in the presence of H2O2 or thiols such as dithiothreitol (DTT). For HRP-based ELISA applications, this results in at least two-fold greater sensitivity compared to Amplex Red.

Benefits of the Amplex UltraRed reagent over Amplex Red:

  • Brighter signals and enhanced sensitivity for HRP activity and H2O2
  • Less sensitive to acidic pH
  • Improved stability in the presence of H2O2 or thiols such as DTT

The Amplex Red and Amplex UltraRed reaction products resorufin and Amplex UltroxRed have high extinction coefficients so they can be measured either fluorometrically or spectrophotometrically with absorbance at ∼560 nm, providing flexibility in detection of H2O2, various enzyme assays, and HRP-based ELISAs.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C).
Detection Method Fluorescence
For Use With (Application) Hydrogen Peroxidase Assay
For Use With (Equipment) Fluorometer, Microplate Reader
Product Type Red Hydrogen Peroxide
Dye Type Amplex Red Reagent
Form Solid
Product Line Amplex
Quantity 5 mg
Shipping Condition Room Temperature
Substrate HRP (Horseradish Peroxidase) Substrate
Substrate Properties Chemical substrate
Validated Application Cell Analysis
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.