Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Click-iT™ EdU Alexa Fluor™ 488 HCS Assay
GREENER_CHOICE

Catalog No. p-4920986
Encompass_Preferred
Click to view available options
Quantity:
10 x 96 well plates
2 x 96 well plates

Includes

The kit contains sufficient material for two 96-well plates.

For high-content imaging applications

The Click-iT EdU Alexa Fluor 488 HCS Assay is a superior alternative to traditional proliferation assays that is optimized for high-content imaging applications. In this assay the modified thymidine analogue EdU is efficiently incorporated into newly synthesized DNA and fluorescently labeled with a bright, photostable Alexa Fluor dye in a fast, highly specific click reaction. This fluorescent labeling of proliferating cells is accurate and compatible with antibody methods due to the mild click protocol. Fast, highly specific click reaction. The most accurate proliferation-detection methods are based on the incorporation and measurement of nucleoside analogues in newly synthesized DNA, with bromodeoxyuridine (BrdU) a commonly used analogue. BrdU-labeled DNA is quantitated using anti-BrdU antibodies following DNA denaturation by harsh methods (HCl, heat, or enzymes) to expose the BrdU molecules. This step is time consuming and difficult to perform consistently. The harsh treatment can also adversely affect sample integrity and quality, which makes co-staining with other antibodies challenging.

  • Simple: works the first time, every time, in less time
  • Efficient: no denaturation steps or harsh treatment required
  • Gentle on samples: better preservation of cell morphology, antigen structure, and DNA integrity
  • Consistent: not dependent on variable antibody lots for detection
  • Less hazardous

Superior Proliferation Methodology
The Click-iT EdU Alexa Fluor 488 HCS Assay provides a superior alternative to BrdU assays for measuring cell proliferation. EdU (5-ethynyl-2'-deoxyuridine) is a nucleoside analog of thymidine and is incorporated into DNA during active DNA synthesis. With Click-iT EdU, mild fixation and detergent permeabilization is sufficient for the small molecule-based Click-iT EdU detection reagent to gain access to the DNA. As a consequence, the Click-iT EdU HCS assay is not only easy to use, but more accurate and compatible with cell cycle analysis and other intracellular or extracellular targets for truly content-rich results.

  • The Click-iTassay can be used on cells in culture or in vivo following administration of EdU by feeding or injection methods.
  • The Click-iTassay can be used with BrdU in dual pulse experiments by using the ant-BrdU (clone MoBu-1) antibody, which does not cross react with EdU.
  • The Click-iTtechnology is compatible with immunohistochemical, immunocytochemical, and fluorescent dyes that are fixation tolerant or designed for fixed-cell labeling.

Cell Analysis, Cell Cycle, Cell Proliferation, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Replication and Repair, High-Content Screening (HCS)

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Green
Content And Storage The kit contains sufficient material for 2 96-well plates. Store at 2–6°C, dessicate and protect from light.
Detection Method Fluorescence
For Use With (Application) High-content Screening (HCS)
For Use With (Equipment) High Content Analysis Instrument
Product Type HCS Assay
Dye Type Alexa Fluor™ 488
Emission Visible
Format 96-well plate
Green Features Less hazardous
Product Line Alexa Fluor, Click-iT, Molecular Probes
Quantity 2 x 96 well plates
Shipping Condition Room Temperature
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.