Learn More
Invitrogen™ Molecular Probes™ Fura Red™, AM, Cell Permeant Non-distribution product as customer accommodation. Available on GSA/VA Contract for Federal Government customers only.
Visible light-excitable fura-2 analog that offers particular possibilities for ratiometric measurement of Ca2+ in single cells
Manufacturer:  Invitrogen™ F3020
 View more versions of this product
Catalog No. F3020
Add to cart
Description
Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Flow Cytometer, Fluorescence Microscope, Microphotometer
Store in freezer (-5° to -30°C) and protect from light.
500μg
Documents
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only. Not for use in diagnostic procedures.