Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Monastrol
GSA_VAGREENER_CHOICE

Catalog No. p-4785132
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 13-051-0

Supplier: Tocris Bioscience™ 1305/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective inhibitor of mitotic kinesin Eg5

Potent, cell-permeable, small molecule mitosis inhibitor that does not interact with tubulin. Arrests cells in mitosis and specifically inhibits the motility of the mitotic kinesin Eg5, a motor protein required for mitotic spindle formation and maintenance (IC50 = 14 μM).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 254753-54-3
Molecular Formula C14H16N2O3S

Specifications

Quantity 10 mg
Formula Weight 292.35
Purity >98%
Molecular Weight (g/mol) 292.353
Chemical Name or Material Monastrol
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.