Learn More
Description
Specifications
Specifications
| Antigen | Mre11 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 20802516), Immunohistochemistry-Paraffin 10 ug/ml |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Accession No. | NP_005581 |
| Gene Alias | ATLDmeiotic recombination (S. cerevisiae) 11 homolog A, AT-like disease, DNA recombination and repair protein, double-strand break repair protein MRE11A, endo/exonuclease Mre11, HNGS1, Meiotic recombination 11 homolog 1, Meiotic recombination 11 homolog A, MRE11 homolog 1, MRE11 homolog A, MRE11 meiotic recombination 11 homolog A (S. cerevisiae), MRE11B |
| Gene Symbols | MRE11 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
