Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRP1 Antibody (IU5C1) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | MRP1 |
---|---|
Clone | IU5C1 |
Dilution | Western Blot 1:250 - 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11057131T
![]() |
Novus Biologicals
NB11057131SS |
0.025 mL |
Each for $206.50
|
|
|||||
NB11057131
![]() |
Novus Biologicals
NB11057131 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
MRP1 Monoclonal specifically detects MRP1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MRP1 | |
Western Blot 1:250 - 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence reported in scientific literature, Immunohistochemistry-Paraffin 1:200 | |
Monoclonal | |
Ascites | |
RUO | |
Human, Mouse | |
P33527 | |
4363 | |
A recombinant peptide corresponding to residues 1-33 (N-terminal: SMALRGFCSADGSDPLWDWNVTWNTSNPDFTKCF) of human MRP1. [Swiss-Prot# P33527] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
172 kDa |
IU5C1 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
ABC Transporters, Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Plasma Membrane Markers, Signal Transduction | |
PBS with 0.1% Sodium Azide | |
ABC29, ABCC, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
ABCC1 | |
IgG1 | |
Protein G purified | |
Human and mouse. Immunogen has 96% homology to rat, primate, and feline, 88% homology to chicken, 87% homology to bovine, 72% homology to Drosophila melanogaster. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title