Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MRPL49 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | MRPL49 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP16893220
![]() |
Novus Biologicals
NBP16893220UL |
20 μL |
Each for $206.00
|
|
|||||
NBP168932
![]() |
Novus Biologicals
NBP168932 |
100 μL |
Each for $487.50
|
|
|||||
Description
MRPL49 Polyclonal specifically detects MRPL49 in Human samples. It is validated for Western Blot.Specifications
MRPL49 | |
Polyclonal | |
Rabbit | |
Stem Cell Markers | |
C11orf4, L49mt39S ribosomal protein L49, mitochondrial, mitochondrial ribosomal protein L49, MRP-L49, Neighbor of FAU, next to FAU, NOF1chromosome 11 open reading frame 4, NOFMGC10656, Protein NOF1 | |
MRPL49 | |
IgG | |
19 kDa |
Western Blot | |
Unconjugated | |
RUO | |
Q13405 | |
740 | |
Synthetic peptides corresponding to MRPL49 (mitochondrial ribosomal protein L49) The peptide sequence was selected from the N terminal of MRPL49. Peptide sequence ESVDEYQFVERLLPATRIPDPPKHEHYPTPSGWQPPRDPPPNLPYFVRRS. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title