Learn More
MSA™ Airline Respirator Manifolds
Compressed airline manifolds help ensure proper air supply from a single air source.
$744.26 - $1242.37
Specifications
For Use With (Equipment) MSA Airline Respirators
Includes Automatic shut-off
Product Type Manifold Assembly
View More Specs
Products 4
Catalog Number Mfr. No. Connection Type Price Quantity & Availability  
Catalog Number Mfr. No. Connection Type Price Quantity & Availability  
18-139-767
View Documents
MSA™
84416
Foster quick-disconnect, two-outlets
Each for $744.26
Only null left
Add to Cart
18-139-760
View Documents
MSA™
486053
Snap-Tite™ quick-disconnect, two-outlets
Each for $744.26
Only null left
Add to Cart
18-139-753
View Documents
MSA™
47370
Foster quick-disconnect, four-outlets
Each for $1,242.37
Only null left
Add to Cart
18-139-762
View Documents
MSA™
93931
Snap-Tite™ quick-disconnect, four-outlets
Each for $1,190.29
Only null left
Add to Cart
Description
Description
  • Enhance long-duration respiratory protection
  • Ensure proper pressure and air quality
  • Regulator reduces air system compressor pressure and maintains desired operating pressure
  • Manifolds feature quick-disconnect assemblies and automatic shut-off outlets
Specifications
Specifications
MSA Airline Respirators
Automatic shut-off
Manifold Assembly
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.