Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ 4X Multigas Detector

Catalog No. 17200060 Shop All MSA Products
Change view
Click to view available options
Resolution:
O2: 0.1%, CO: 1ppm, LEL: 1%
O2: 0.1%, LEL: 1%
O2: 0.1%, LEL: 1%, CO: 1%
Dimensions (L x W x H):
4 x 3 x 4.4 in. (10.16 x 7.62 x 10.1cm)
7 x 3 x 4.4 in. (18 x 7.62 x 10.1cm)
Weight (English):
19.4 oz.
23.84 oz.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Resolution Dimensions (L x W x H) Weight (English)
17-200-060 O2: 0.1%, CO: 1ppm, LEL: 1% 4 x 3 x 4.4 in. (10.16 x 7.62 x 10.1cm) 19.4 oz.
19-100-811 O2: 0.1%, LEL: 1%, CO: 1% 7 x 3 x 4.4 in. (18 x 7.62 x 10.1cm) 23.84 oz.
17-200-061 O2: 0.1%, LEL: 1% 4 x 3 x 4.4 in. (10.16 x 7.62 x 10.1cm) 19.4 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-200-060 Supplier MSA™ Supplier No. 10110445

Second generation of the ALTAIR 4 Multigas Detector

Driven by MSA XCell™ sensor technology.

  • Personal four-gas monitor—all models detect LEL and O2; selected models detect CO or CO plus H2S
  • Extreme durability—survives 10 ft.+ drop test
  • Functionality and ease of use—both operate the same way
  • IP67 Rated—can be submerged in water
  • MotionAlert™ feature activates if unit does not detect motion for 30 seconds
  • InstantAlert™ feature allows users to manually alert others to a potentially hazardous situation
  • Galaxy™ system compatible for calibration and record keeping
  • QuickCheck™ Test Station for fast, easy bump testing available (sold separately)
  • Datalogging and MSA Link™ software ready

Improved features

  • New MSA XCell Sensors—longer life and higher performance
  • Longer runtime—24-hour continuous runtime
  • End-of-sensor-life warning—heart icon when nearing end of life
  • Improved buttons—softer, larger, easier to press
  • White backlight—improved readability
MSA XCell Sensor Technology
  • Typical sensor life: 4 years
  • Typical response time: ≤15 seconds faster response (clear time, bump test)
  • Embedded microelectronics provide greater stability and performance; each sensor has its own microprocessor and digital output
  • Leak-tight—laser-welded sensor sealing for increased durability

Order Info

For information on ratings and certifications, contact your Fisher Safety Customer Service Representative.

Certifications

UL and CSA approved. Also meets various international requirements. Contact your Fisher Safety Customer Service Representative for details.

Warranty and Services

Three years

Specifications

Alarm Types Audible, visual, vibrating
Battery Life 24 hr.
Components Measured Hydrogen Sulfide, LEL, Oxygen
Dimensions (L x W x H) 4 x 3 x 4.4 in. (10.16 x 7.62 x 10.1cm)
Height (English) 4.4 in.
Height (Metric) 10.1 cm
IP Rating IP67
Length (English) 4 in.
Length (Metric) 10.16 cm
Resolution O2: 0.1%, CO: 1ppm, LEL: 1%
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 3 Years
Weight (English) 19.4 oz.
Weight (Metric) 540g
Width (English) 3 in.
Width (Metric) 7.62 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.