Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA™ ALTAIR™ 5X Multigas Detector

Catalog No. 17200066 Shop All MSA Products
Change view
Click to view available options
Components Measured:
Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen
LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide
LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide
Type:
Multigas Detector: Deluxe Kit
Multigas Detector: Industrial Kit
Multigas Detector: Standard Kit
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Components Measured Type
17-200-066 LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide Multigas Detector: Deluxe Kit
17-200-064 LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide Multigas Detector: Industrial Kit
17-200-062 Carbon Monoxide, Hydrogen Sulfide, LEL, Oxygen Multigas Detector: Standard Kit
17-200-063 LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide Multigas Detector: Standard Kit
17-200-065 LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide Multigas Detector: Industrial Kit
17-200-067 LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide, Sulfur Dioxide Multigas Detector: Deluxe Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 17-200-066 Supplier MSA™ Supplier No. 10116928/DUP

Includes

  • Detector Industrial Kit includes detector with monochrome display, integral pump, 10
  • ft. sampling line and 1 ft. probe. Detector Deluxe Kit includes detector with color display, integral pump, 10 ft. sampling line and 1 ft. probe

Monitors up to six gases simultaneously — ideal for confined space monitoring

New XCell™ sensor technology doubles the typical life span of sensors, offers faster response and span calibration times and provides plug-and-play capabilities for easy reconfiguration. Sensors available for SO2, Cl2 and NH3 for expanded monitoring applications.

  • Sensor response and clear times under 15 seconds for most common sensor configurations
  • End of sensor life warning provides advanced notice to user, reducing service outages
  • MotionAlert activates if user becomes disabled and motionless and provides user's location
  • InstantAlert enables users to manually alert others to potentially hazardous situations
  • Span calibration time of 60 seconds for most common sensor configurations
  • 24-hour bump checkmark; bump test under 15 seconds for most common configurations
  • Robust integral pump for consistent flow without externally attached components
  • 18 language options
  • Durable, rubberized housing for secure grip
  • Lithium-ion battery lasts up to 20 hours
  • High-contrast monochrome or color display
  • Fully compatible with Galaxy™ Automated Test System and MSA Link™ software

Order Info

L x W x D: 6.69 x 3.49 x 1.79 in.. Weight: 1 lb.

Compliance

IP65 rated. Approvals: Class I, Division 1, Groups A, B, C and D; Class II Division 1 Groups E, F and G; Class III Division 1

Warranty and Services

Three years on detector, CO, H2S, O2, LEL and SO2 sensors.

Specifications

Alarm Types Audible, visual, vibrating
Battery Life 20 hr.
Components Measured LEL, Oxygen, Carbon Monoxide, Hydrogen Sulfide
Dimensions (L x W x H) 3.5 x 3.8 x 3.75 in. (8.89 x 9.65 x 9.53cm)
Type Multigas Detector: Deluxe Kit
Height (English) 3.75 in.
Height (Metric) 9.53 cm
IP Rating IP65
Length (English) 3.5 in.
Length (Metric) 8.89 cm
Resolution O2: 0.1%, H2S: 1ppm, CO: 1ppm, LEL: 1%
Temperature Range (English) -4°F to +122°F
Temperature Range (Metric) -20°C to +50°C
Warranty 3 Years
Weight (English) 27.84 oz.
Weight (Metric) 790g
Width (English) 3.8 in.
Width (Metric) 9.65 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.