Learn More
MSA™ Calibration Kits and Gases
Catalog No. p-27483 Shop All MSA Products
Change view
Click to view available options
Type:
Calibration Kit
Regulator
For Use With (Equipment):
Model R Calibration Check Kits
Model R Gas Cylinders
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
18-999-3343 Regulator Model R Calibration Check Kits
19-120-1216 Calibration Kit -
18-999-3237 Calibration Kit Model R Gas Cylinders
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-999-3343 Supplier MSA™ Supplier No. 459948
Only null left
Add to Cart
Add to Cart
Offers quick and convenient method of checking response of MSA gas sensors and gas detection instrumentation
  • Kits contains necessary calibration components to calibrate equipment
  • Housed in lightweight carrying case for easy transport
Specifications
Description For checking response of MSA gas sensors and gas detection instrumentation
For Use With (Equipment) Model R Calibration Check Kits
Type Regulator
Flow Rate 1.5
Includes Regulator